
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in sheep

Synonym(s):
SAM
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Technique
Showing 1-2 of 2