Skip to Content
Merck
Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody, Mouse monoclonal clone MAPK-YT, purified from hybridoma cell culture

Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody, Mouse monoclonal

Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody, Mouse monoclonal clone MAPK-YT, purified from hybridoma cell culture
Synonym(s):
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2)

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing