![Anti-DC-SIGN antibody produced in rabbit affinity isolated antibody](/deepweb/assets/sigmaaldrich/product/images/277/707/381bb0bc-3415-4aad-be74-92d0b175e1f9/640/381bb0bc-3415-4aad-be74-92d0b175e1f9.jpg)
Anti-DC-SIGN antibody produced in rabbit
![Anti-DC-SIGN antibody produced in rabbit affinity isolated antibody](/deepweb/assets/sigmaaldrich/product/images/277/707/381bb0bc-3415-4aad-be74-92d0b175e1f9/640/381bb0bc-3415-4aad-be74-92d0b175e1f9.jpg)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing