
Amiloride Related Compound A

Synonym(s):
Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
Empirical Formula (Hill Notation):
C6H7ClN4O2
CAS Number:
Molecular Weight:
202.60
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2