
2-(6-Methoxynaphthalen-2-yl)acetic acid

Synonym(s):
6-Methoxy-2-naphthaleneacetic acid, 6-Methoxy-2-naphthylacetic acid
Empirical Formula (Hill Notation):
C13H12O3
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing