
2,2′,5,5′-Tetrachlorobiphenyl (IUPAC No. 52)

Synonym(s):
2,2′,5,5′-Tetrachlorobiphenyl
Empirical Formula (Hill Notation):
C12H6Cl4
CAS Number:
Molecular Weight:
291.99
Products
Formula Weight
Manufacturer
Agency
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing