Skip to Content
Merck
1-Hydroxy-7-azabenzotriazole solution ~0.6 M in DMF

1-Hydroxy-7-azabenzotriazole solution

1-Hydroxy-7-azabenzotriazole solution ~0.6 M in DMF
Synonym(s):
3H-[1,2,3]-Triazolo[4,5-b]pyridin-3-ol, HOAt
Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11

Products

Brand
Formula Weight
Physical Form
Application
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing