
1-Cyano-4-dimethylaminopyridinium tetrafluoroborate

Synonym(s):
CDAP
Empirical Formula (Hill Notation):
C8H10BF4N3
CAS Number:
Molecular Weight:
234.99
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing