Merck
  • Novel thermoresponsive assemblies of co-grafted natural and synthetic polymers for water purification.
Novel thermoresponsive assemblies of co-grafted natural and synthetic polymers for water purification.
Water science and technology : a journal of the International Association on Water Pollution Research (2017-03-09)
Joginder Singh Paneysar, Stephen Barton, Sudeshna Chandra, Premlata Ambre, Evans Coutinho
ABSTRACT
Water contamination and its purification are a global problem. The current approach to purify water is reduction of impurities to acceptable levels. One of the ways to achieve this is by use of water-soluble polymers that extract organic and metallic contaminants, from water. This paper presents a blend of composite polymers that eliminates both the contaminants simultaneously by the principle of adsorption at lower critical solution temperature. These composite polymers have been synthesized by grafting poly(N,N-diethylacrylamide), poly(N-isopropylacrylamide) and poly(N-vinylcaprolactam) on-to the natural polymer chitosan or its derivatives, giving smart graft polymeric assemblies (GPAs). One of the graft polymers, GPA-2, exhibits excellent adsorption properties able to remove metal ions like cadmium, cobalt, copper, lead, iron and also organic impurities like chlorophenol and phthalic anhydride. Studies reveal that 6 mg/ml GPA-2 is able to effect a 100% removal of organic impurities - chlorophenol (50 ppm) and phthalic anhydride (70 ppm) - from water, while complete removal of the heavy metal ions (Cu
Sigma-Aldrich
N,N-Diethylacrylamide, contains <200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
N-Vinylcaprolactam, 98%, stabilized