Merck
  • Secalonic Acid-D Represses HIF1α/VEGF-Mediated Angiogenesis by Regulating the Akt/mTOR/p70S6K Signaling Cascade.
Secalonic Acid-D Represses HIF1α/VEGF-Mediated Angiogenesis by Regulating the Akt/mTOR/p70S6K Signaling Cascade.
Cancer research (2015-05-16)
Santosh Kumar Guru, Anup Singh Pathania, Suresh Kumar, Deshidi Ramesh, Manjeet Kumar, Satiander Rana, Ajay Kumar, Fayaz Malik, P R Sharma, B K Chandan, Sundeep Jaglan, J P Sharma, Bhahwal Ali Shah, Sheikh Abdullah Tasduq, Surrinder K Lattoo, Abdul Faruk, A K Saxena, R A Vishwakarma, Shashi Bhushan
ABSTRACT
Tumor angiogenesis is a validated target for therapeutic intervention, but agents that are more disease selective are needed. Here, we report the isolation of secalonic acid-D (SAD), a mycotoxin from a novel source that exhibits potent antiangiogenic antitumor activity. SAD inhibited multiple HIF1α/VEGF-arbitrated angiogenesis dynamics as scored in human umbilical vascular endothelial cells and human MCF-7 breast tumor xenografts. Similarly, SAD suppressed VEGF-induced microvessel sprouting from rat aortic ring and blood vessel formation in the Matrigel plug assay in C57/BL6J mice. Under normoxic or hypoxic conditions, SAD inhibited cell survival through the Akt/mTOR/p70S6K pathway, with attendant effects on key proangiogenesis factors, including HIF1α, VEGFR, and MMP-2/MMP-9. These effects were reversed by cotreatment with the Akt inhibitors perifosine and GSK69069 or by the addition of neutralizing VEGF antibodies. The apoptotic properties of SAD were determined to be both extrinsic and intrinsic in nature, whereas the cell-cycle inhibitory effects were mediated by altering the level of key G1-S transition-phase proteins. In experimental mouse models of breast cancer, SAD dosing produced no apparent toxicities (either orally or intraperitoneal) at levels that yielded antitumor effects. Taken together, our findings offered a preclinical validation and mechanistic definition of the antiangiogenic activity of a novel mycotoxin, with potential application as a cancer-selective therapeutic agent.
Product Description
PC-3 Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 90112714, human prostate, Epithelial
Sigma-Aldrich
QCM Chemotaxis Cell Migration Assay, 24-well (8 µm), fluorimetric, The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with fluorescent detection.