Merck
  • Inhibition of KIF14 Suppresses Tumor Cell Growth and Promotes Apoptosis in Human Glioblastoma.
Inhibition of KIF14 Suppresses Tumor Cell Growth and Promotes Apoptosis in Human Glioblastoma.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-11-05)
Wei Huang, JunYu Wang, Danfeng Zhang, Wen Chen, Lijun Hou, Xiaojun Wu, Yicheng Lu
ABSTRACT
The mitotic kinesin superfamily protein KIF14 is essential for cytokinesis and chromosome segregation, and increased KIF14 expression is related to a variety of human cancers. However, the role of KIF14 in the development and malignant progression of astrocytomas and the underlying mechanisms remain unclear. The present study examined the relation between KIF14 and the pathogenesis of malignant astrocytoma. The role of KIF14 in astrocytoma development and progression was investigated by analyzing KIF14 expression using SYBR Green quantitative real-time RT-PCR, western blotting and immunohistochemistry in human astrocytoma and normal brain tissues. KIF14 expression was higher in astrocytoma samples, and was positively correlated with pathological grade and proliferative activity indicated by Ki-67 staining. SiRNA knockdown of KIF14 inhibited tumor growth in vitro and in vivo, attenuated anchorage-independent growth, and induced G2/M phase arrest, cytokinesis failure and apoptosis in glioblastoma cell lines in association with decreased AKT phosphorylation and activity. The upregulation of KIF14 in astrocytoma is associated with disease severity, and suppression of KIF14 inhibits cell proliferation and induces apoptosis through a mechanism involving the inactivation of AKT signaling, suggesting that KIF14 plays an important role in astrocytoma tumorigenesis and could be a promising molecular target for anticancer therapy.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human KIF14
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Kif14
Sigma-Aldrich
DAPI, for nucleic acid staining