Merck
  • Chronological and replicative lifespan of polyploid Saccharomyces cerevisiae (syn. S. pastorianus).
Chronological and replicative lifespan of polyploid Saccharomyces cerevisiae (syn. S. pastorianus).
FEMS yeast research (2003-04-19)
Dawn L Maskell, Alan I Kennedy, Jeff A Hodgson, Katherine A Smart
ABSTRACT
Chronological lifespan may be defined as the result of accumulation of irreversible damage to intracellular components during extended stationary phase, compromising cellular integrity and leading to death and autolysis. In contrast, replicative lifespan relates to the number of divisions an individual cell has undertaken before entering a non-replicative state termed senescence, leading to cell death and autolysis. Both forms of lifespan have been considered to represent models of ageing in higher eukaryotes, yet the relation between chronologically and replicatively aged populations has not been investigated. In this study both forms of lifespan have been investigated in Saccharomyces cerevisiae (Syn. S. pastorianus) to establish the relationship between chronological and replicative ageing.
Sigma-Aldrich
Methylene Violet (Bernthsen), certified by the Biological Stain Commission, Dye content ≥65 %