Merck
  • Estimation of cholinesterase inhibitory and antioxidant effects of the leaf extracts of Anatolian Ficus carica var. domestica and their total phenol and flavonoid contents.
Estimation of cholinesterase inhibitory and antioxidant effects of the leaf extracts of Anatolian Ficus carica var. domestica and their total phenol and flavonoid contents.
Natural product communications (2011-04-13)
Ilkay Erdogan Orhan, Osman Ustün, Bilge Sener
ABSTRACT
Ficus carica var. domestica Tsch. & Rav. (common fig) is widely grown in Turkey and exported for its edible fruits. In this study, the n-hexane, chloroform, acetone, methanol, n-butanol, and water extracts of the leaves of F. carica var. domestica were screened for their cholinesterase inhibitory and antioxidant activities. Cholinesterase inhibition against acetyl- (AChE) and butyrylcholinesterase (BChE) was measured by the spectrophotometric method of Ellman at concentrations of 25, 50, and 100 microg/mL., while antioxidant activity was tested using three in vitro methods; 2,2-diphenyl-1-picrylhydrazyl (DPPH) radical scavenging activity, metal-chelation capacity, and ferric-reducing antioxidant power (FRAP). Total phenol and flavonoid contents of the extracts were determined spectrophotometrically. Our results revealed that the n-hexane and acetone extracts exerted a notable inhibition against both AChE (62.9 +/- 0.9% and 50.8 +/- 2.1%, respectively) and BChE (76.9 +/- 2.2% and 45.6 +/- 1.3%, respectively). However, they had low activity in the antioxidant tests. The chloroform extract was found to be the richest in total flavonoid content (252.5 +/- 1.1 mg/g quercetin equivalent), while the n-butanol extract had the highest total phenol amount (85.9 +/- 3.2 mg/g extract gallic acid equivalent).
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p′-disulfonic acid monosodium salt hydrate, 97%
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, Indicator ligand
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, BioXtra
Supelco
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, for spectrophotometric det. of Fe, 97.0%