Merck
  • CDK7 controls E2F- and MYC-driven proliferative and metabolic vulnerabilities in multiple myeloma.
CDK7 controls E2F- and MYC-driven proliferative and metabolic vulnerabilities in multiple myeloma.
Blood (2023-03-07)
Yao Yao, Jessica Fong Ng, Woojun Daniel Park, Mehmet Samur, Eugenio Morelli, Jessica Encinas Mayoral, Zuzana Chyra, Yan Xu, Sanika Derebail, Charles Epstein, Behnam Nabet, Marta Chesi, Nathanael S Gray, Richard A Young, Nicholas Kwiatkowski, Constantine Mitsiades, Kenneth C Anderson, Charles Y Lin, Nikhil C Munshi, Mariateresa Fulciniti
ABSTRACT
Therapeutic targeting of CDK7 has proven beneficial in preclinical studies, yet the off-target effects of currently available CDK7 inhibitors make it difficult to pinpoint the exact mechanisms behind MM cell death mediated by CDK7 inhibition. Here, we show that CDK7 expression positively correlates with E2F and MYC transcriptional programs in cells from patients with multiple myeloma (MM); its selective targeting counteracts E2F activity via perturbation of the cyclin-dependent kinases/Rb axis and impairs MYC-regulated metabolic gene signatures translating into defects in glycolysis and reduced levels of lactate production in MM cells. CDK7 inhibition using the covalent small-molecule inhibitor YKL-5-124 elicits a strong therapeutic response with minimal effects on normal cells, and causes in vivo tumor regression, increasing survival in several mouse models of MM including a genetically engineered mouse model of MYC-dependent MM. Through its role as a critical cofactor and regulator of MYC and E2F activity, CDK7 is therefore a master regulator of oncogenic cellular programs supporting MM growth and survival, and a valuable therapeutic target providing rationale for development of YKL-5-124 for clinical use.
Sigma-Aldrich
ChIPAb+ E2F-1 - ChIP Validated Antibody and Primer Set, from mouse