Merck
  • Autophagy in T cells from aged donors is maintained by spermidine and correlates with function and vaccine responses.
Autophagy in T cells from aged donors is maintained by spermidine and correlates with function and vaccine responses.
eLife (2020-12-16)
Ghada Alsaleh, Isabel Panse, Leo Swadling, Hanlin Zhang, Felix Clemens Richter, Alain Meyer, Janet Lord, Eleanor Barnes, Paul Klenerman, Christopher Green, Anna Katharina Simon
ABSTRACT
Vaccines are powerful tools to develop immune memory to infectious diseases and prevent excess mortality. In older adults, however vaccines are generally less efficacious and the molecular mechanisms that underpin this remain largely unknown. Autophagy, a process known to prevent aging, is critical for the maintenance of immune memory in mice. Here, we show that autophagy is specifically induced in vaccine-induced antigen-specific CD8+ T cells in healthy human volunteers. In addition, reduced IFNγ secretion by RSV-induced T cells in older vaccinees correlates with low autophagy levels. We demonstrate that levels of the endogenous autophagy-inducing metabolite spermidine fall in human T cells with age. Spermidine supplementation in T cells from old donors recovers their autophagy level and function, similar to young donors' cells, in which spermidine biosynthesis has been inhibited. Finally, our data show that endogenous spermidine maintains autophagy via the translation factor eIF5A and transcription factor TFEB. In summary, we have provided evidence for the importance of autophagy in vaccine immunogenicity in older humans and uncovered two novel drug targets that may increase vaccination efficiency in the aging context.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Deoxyhypusine Synthase Inhibitor, GC7, The Deoxyhypusine Synthase Inhibitor, GC7, also referenced under CAS 150333-69-0, controls the biological activity of Deoxyhypusine Synthase.
Sigma-Aldrich
Anti-LC3 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution