Merck
  • Regulation of neural gene transcription by optogenetic inhibition of the RE1-silencing transcription factor.
Regulation of neural gene transcription by optogenetic inhibition of the RE1-silencing transcription factor.
Proceedings of the National Academy of Sciences of the United States of America (2015-12-25)
Francesco Paonessa, Stefania Criscuolo, Silvio Sacchetti, Davide Amoroso, Helena Scarongella, Federico Pecoraro Bisogni, Emanuele Carminati, Giacomo Pruzzo, Luca Maragliano, Fabrizia Cesca, Fabio Benfenati
ABSTRACT
Optogenetics provides new ways to activate gene transcription; however, no attempts have been made as yet to modulate mammalian transcription factors. We report the light-mediated regulation of the repressor element 1 (RE1)-silencing transcription factor (REST), a master regulator of neural genes. To tune REST activity, we selected two protein domains that impair REST-DNA binding or recruitment of the cofactor mSin3a. Computational modeling guided the fusion of the inhibitory domains to the light-sensitive Avena sativa light-oxygen-voltage-sensing (LOV) 2-phototrophin 1 (AsLOV2). By expressing AsLOV2 chimeras in Neuro2a cells, we achieved light-dependent modulation of REST target genes that was associated with an improved neural differentiation. In primary neurons, light-mediated REST inhibition increased Na(+)-channel 1.2 and brain-derived neurotrophic factor transcription and boosted Na(+) currents and neuronal firing. This optogenetic approach allows the coordinated expression of a cluster of genes impinging on neuronal activity, providing a tool for studying neuronal physiology and correcting gene expression changes taking place in brain diseases.
Sigma-Aldrich
Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks