Merck
  • Expeditious synthesis of tri- and tetrahydroxyazepanes from D-(-)-quinic acid as potent glycosidase inhibitors.
Expeditious synthesis of tri- and tetrahydroxyazepanes from D-(-)-quinic acid as potent glycosidase inhibitors.
The Journal of organic chemistry (2007-05-08)
Tzenge-Lien Shih, Ru-Ying Yang, Shiou-Ting Li, Cheng-Fan Chiang, Chun-Hung Lin
ABSTRACT
Several new stereoisomers of 3,4,6-trihydroxyazepanes and 7-hydroxymethyl-3,4,5-trihydroxyazepanes as well as known 3,4,5-trihydroxyazepanes were synthesized as potent glycosidase inhibitors from D-(-)-quinic acid in an efficient manner. The key step employs dihydroxylation of protected chiral 1,4,5-cyclohex-2-enetriols under RuCl3/NaIO4/phosphate buffer (pH 7) condition, followed by reductive amino cyclization. We found the choice of an appropriate protecting group to C1-OH of chiral 1,4,5-cyclohex-2-enetriols would increase the yields of cyclization. The preliminary biological data indicate some of these azepanes possess potent inhibition against alpha-mannosidase and alpha-fucosidase.
Sigma-Aldrich
D-(−)-Quinic acid, 98%