Merck
  • MRP1 modulators synergize with buthionine sulfoximine to exploit collateral sensitivity and selectively kill MRP1-expressing cancer cells.
MRP1 modulators synergize with buthionine sulfoximine to exploit collateral sensitivity and selectively kill MRP1-expressing cancer cells.
Biochemical pharmacology (2019-07-16)
Christine C Gana, Kimberley M Hanssen, Denise M T Yu, Claudia L Flemming, Madeleine S Wheatley, Gwenaëlle Conseil, Susan P C Cole, Murray D Norris, Michelle Haber, Jamie I Fletcher
ABSTRACT
Members of the ABC transporter family, particularly P-glycoprotein (P-gp, ABCB1), breast cancer resistance protein (BCRP, ABCG2) and multidrug resistance protein 1 (MRP1, ABCC1) are well characterized mediators of multidrug resistance, however their pharmacological inhibition has so far failed as a clinical strategy. Harnessing collateral sensitivity, a form of synthetic lethality where cells with acquired multidrug resistance exhibit hypersensitivity to unrelated agents, may be an alternative approach to targeting multidrug resistant tumour cells. We characterized a novel small molecule modulator that selectively enhanced MRP1-dependent efflux of reduced glutathione (GSH), an endogenous MRP1 substrate. Using cell lines expressing high levels of endogenous MRP1 from three difficult to treat cancer types-lung cancer, ovarian cancer and high-risk neuroblastoma-we showed that the MRP1 modulator substantially lowered intracellular GSH levels as a single agent. The effect was on-target, as MRP1 knockdown abolished GSH depletion. The MRP1 modulator was synergistic with the GSH synthesis inhibitor buthionine sulfoximine (BSO), with the combination exhausting intracellular GSH, increasing intracellular reactive oxygen species (ROS) and abolishing clonogenic capacity. Clonogenicity was rescued by the ROS scavenger N-acetylcysteine, implicating GSH depletion in the effect. The MRP1 modulator in combination with BSO also strongly sensitized cancer cells to MRP1-substrate chemotherapeutic agents, particularly arsenic trioxide, and was more effective than either the MRP1 modulator or BSO alone. GSH-depleting MRP1 modulators may therefore provide an enhanced therapeutic window to treat chemo-resistant MRP1-overexpressing pediatric and adult cancers.
Sigma-Aldrich
2-Vinylpyridine, 97%