
Yohimbine for system suitability

Synonym(s):
Yohimbine hydrochloride, 17-Hydroxyyohimban-16-carboxylic acid methyl ester hydrochloride
Empirical Formula (Hill Notation):
C21H26N2O3 · HCl
CAS Number:
Molecular Weight:
390.90
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing