
TissueFab® Sodium Persulfate Solution

Synonym(s):
Sodium persulfate, Sodium peroxodisulfate
Linear Formula:
Na2S2O8
CAS Number:
Molecular Weight:
238.10
Products
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing