Skip to Content
Merck
Thiamine monophosphate chloride dihydrate

Thiamine monophosphate chloride dihydrate

Thiamine monophosphate chloride dihydrate
Synonym(s):
Aneurine monophosphate chloride
Empirical Formula (Hill Notation):
C12H18ClN4O4PS · 2H2O
CAS Number:
Molecular Weight:
416.82
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing