
Rivastigmine tartrate R-isomer

Synonym(s):
(R)-Rivastigmine tartrate, (R)-3-[1-(Dimethylamino)ethyl]phenyl ethylmethylcarbamate hydrogen tartrate
Empirical Formula (Hill Notation):
C14H22N2O2 · C4H6O6
CAS Number:
Molecular Weight:
400.42
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2