
(R)-(+)-2′-Amino-1,1′-binaphthalen-2-ol

Synonym(s):
(R)-(+)-2-Amino-2′-hydroxy-1,1′-binaphthalene, (R)-NOBIN
Empirical Formula (Hill Notation):
C20H15NO
CAS Number:
Molecular Weight:
285.34
Products
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing