
L-Threonine-13C4

Linear Formula:
13CH313CH(OH)13CH(NH2)13CO2H
CAS Number:
Molecular Weight:
123.09
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing