
λ-Cyhalothrin

Empirical Formula (Hill Notation):
C23H19ClF3NO3
Molecular Weight:
449.85
EC Number:
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2