
Isoflurane Related Compound A

Synonym(s):
Chlorodifluoromethyl 1-chloro-2,2,2-trifluoroethyl ether, 1-Chloro-2,2,2-trifluoroethyl chlorodifluoromethyl ether
Empirical Formula (Hill Notation):
C3HCl2F5O
CAS Number:
Molecular Weight:
218.94
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2