
Isocarboxazid

Synonym(s):
5-Methyl-3-isoxazolecarboxylic acid 2-benzylhydrazide
Empirical Formula (Hill Notation):
C12H13N3O2
CAS Number:
Molecular Weight:
231.25
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2