
Doxapram hydrochloride

Synonym(s):
Doxapram hydrochloride monohydrate, 1-Ethyl-4-[2-(4-morpholinyl)ethyl]-3,3-diphenyl-2-pyrrolidinone monohydrochloride monohydrate
Empirical Formula (Hill Notation):
C24H30N2O2 · HCl · H2O
CAS Number:
Molecular Weight:
432.98
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade