
DMT-2′O-MOE-rMeC(bz) Phosphoramidite

Empirical Formula (Hill Notation):
C50H60N5O10P
CAS Number:
Molecular Weight:
922.01
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing