
Dicloxacillin sodium

Synonym(s):
Dicloxacillin sodium salt monohydrate
Empirical Formula (Hill Notation):
C19H16Cl2N3NaO5S·H2O
CAS Number:
Molecular Weight:
510.32
Products
Product Category
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade