
Clemastine for system suitability

Synonym(s):
Clemastine fumarate salt
Empirical Formula (Hill Notation):
C21H26ClNO · C4H4O4
CAS Number:
Molecular Weight:
459.96
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing