
Chebulinic acid

Synonym(s):
Eutannin, NSC 69862
Empirical Formula (Hill Notation):
C41H32O27
CAS Number:
Molecular Weight:
956.68
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing