Key Documents
ZLXLPLUGP
Milli-Q
RO Regeneration Tool
plug, For water purification systems with two Progard® Packs installed
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
General description
The RO Regeneration Tool/Plug is used along with the RO Regeneration Tool (ZLXLCLPAK) when the system has 2 Progard® Packs installed. RO Regeneration tools are used in place of real Progard® packs whenever a RO cleaner (Cl2 tablets, RO Clean-A and RO Clean-B) is being used.
Application
Used for the pH cleaning of the system using chlorine tablets when 2 Progard® Packs are installed.
Features and Benefits
Genuine accessory for RO Regeneration Tool/Plug - ZLXLP0D01
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany