Merck
  • Direct HPV E6/Myc interactions induce histone modifications, Pol II phosphorylation, and hTERT promoter activation.
Direct HPV E6/Myc interactions induce histone modifications, Pol II phosphorylation, and hTERT promoter activation.
Oncotarget (2017-12-10)
Yiyu Zhang, Aleksandra Dakic, Renxiang Chen, Yuhai Dai, Richard Schlegel, Xuefeng Liu
ABSTRACT
Human Papillomavirus Viruses (HPVs) are associated with the majority of human cervical and anal cancers and 10-30% of head and neck squamous carcinomas. E6 oncoprotein from high risk HPVs interacts with the p53 tumor suppressor protein to facilitate its degradation and increases telomerase activity for extending the life span of host cells. We published previously that the Myc cellular transcription factor associates with the high-risk HPV E6 protein
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Myc
Sigma-Aldrich
10058-F4, ≥98% (HPLC), solid
Sigma-Aldrich
MISSION® esiRNA, targeting human MYC
Millipore
Glutathione−Agarose, lyophilized powder
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution