Merck
  • Potential of biochar filters for onsite sewage treatment: Adsorption and biological degradation of pharmaceuticals in laboratory filters with active, inactive and no biofilm.
Potential of biochar filters for onsite sewage treatment: Adsorption and biological degradation of pharmaceuticals in laboratory filters with active, inactive and no biofilm.
The Science of the total environment (2017-08-30)
Sahar Dalahmeh, Lutz Ahrens, Meritxell Gros, Karin Wiberg, Mikael Pell
ABSTRACT
This study investigated the potential of biochar filters as a replacement or complement for sand filters for removal of pharmaceutically active compounds (PhACs) from wastewater in onsite sewage facilities (OSSF). Specifically, the study investigated the effects of biodegradation, adsorption and a combination of these processes on removal of four model PhACs from wastewater in biochar filters operated under hydraulic loading conditions mimicking those found in onsite infiltration beds. Concentrations and removal of the four PhACs (i.e. carbamazepine, metoprolol, ranitidine and caffeine) were investigated over 22weeks in four treatments: biochar (BC) with active or inactive biofilm (BC-active-biofilm, BC-inactive-biofilm), biochar without biofilm (BC-no-biofilm) and sand with active biofilm (Sand-active-biofilm). The adsorption of carbamazepine was high in BC-no-biofilm (99% removal after 22weeks), while biodegradation was very low in Sand-active-biofilm (7% removal after 22weeks). Removal of carbamazepine in BC-active-biofilm was high and stable over the 22weeks (>98%), showing a significant role of biofilm in filter biogeneration. However, carbamazepine removal declined over time in BC-inactive-biofilm, from 99% in week 13 to 73% in week 22. Metoprolol was poorly degraded in Sand-active-biofilm (37% after 22weeks), while adsorption seemed to be the major pathway for removal of metoprolol in biochar. Ranitidine and caffeine were efficiently removed by either adsorption (97% and 98%, respectively, after 22weeks) or biodegradation (99% and >99%, respectively, after 22weeks). In conclusion, biochar is a promising filter medium for OSSF, especially for persistent PhACs such as carbamazepine and metoprolol.
Product Description
Supelco
Atenolol-d7, analytical standard
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)