Merck
  • Joint Antiangiogenic Effect of ATN-161 and Anti-VEGF Antibody in a Rat Model of Early Wet Age-Related Macular Degeneration.
Joint Antiangiogenic Effect of ATN-161 and Anti-VEGF Antibody in a Rat Model of Early Wet Age-Related Macular Degeneration.
Molecular pharmaceutics (2016-04-19)
Wen-Qiu Wang, Feng-Hua Wang, Wen-Xin Qin, Hai-Yun Liu, Bing Lu, Christopher Chung, Jie Zhu, Qing Gu, William Shi, Cindy Wen, Frances Wu, Kang Zhang, Xiao-Dong Sun
ABSTRACT
The wet form of age-related macular degeneration (AMD) is a leading cause of blindness among elderly Americans and is characterized by abnormal vessel growth, termed choroidal neovascularization (CNV). Integrin α5β1 is a transmembrane receptor that binds matrix macromolecules and proteinases to stimulate angiogenesis. We recently demonstrated that integrin α5β1 plays a critical role in the development of choroidal neovascularization. In this study, we determined the role and underlying mechanisms of integrin α5β1 in angiogenesis in human choroidal endothelial cells and evaluated the antiangiogenic effects of delivering a combination therapy of ATN-161, an integrin α5β1 inhibitor, and an anti-VEGF monoclonal antibody to rats with laser-induced CNV. Vascular endothelial growth factor (VEGF) is a signaling protein that stimulates vasculogenesis and angiogenesis through a pathway that is distinct from the integrin α5β1 signaling pathway. Our results indicate that fibronectin binds to integrin α5β1 and synergizes VEGF-induced angiogenesis via two independent signaling pathways, FN/integrin α5β1/FAK/ERK1/2 and FN/integrin α5β1/FAK/AKT. Integrin α5 knockdown by shRNA inhibits endothelial cell migration, tube formation, and proliferation, while ATN-161 only partially decreases integrin α5 function. Treatment with ATN-161 combined with anti-VEGF antibody showed joint effects in attenuating angiogenesis. In summary, our results provide the first evidence for the mechanisms by which integrin α5β1 is involved in ocular pathological neovascularization in vivo, suggesting that dual inhibition of integrin α5β1 and VEGF may be a promising novel therapeutic strategy for CNV in wet AMD.
Sigma-Aldrich
ATN-161, ≥98% (HPLC)
Pricing and availability is not currently available.