Merck
  • Aminodifluorosulfinium tetrafluoroborate salts as stable and crystalline deoxofluorinating reagents.
Aminodifluorosulfinium tetrafluoroborate salts as stable and crystalline deoxofluorinating reagents.
Organic letters (2009-10-06)
Francis Beaulieu, Louis-Philippe Beauregard, Gabriel Courchesne, Michel Couturier, François LaFlamme, Alexandre L'Heureux
ABSTRACT
Aminodifluorosulfinium tetrafluoroborate salts were found to act as efficient deoxofluorinating reagents when promoted by an exogenous fluoride source and, in most cases, exhibited greater selectivity by providing less elimination byproduct as compared to DAST and Deoxo-Fluor. Aminodifluorosulfinium tetrafluoroborates are easy handled crystalline salts that show enhanced thermal stability over dialkylaminosulfur trifluorides, are storage-stable, and unlike DAST and Deoxo-Fluor do not react violently with water.
Sigma-Aldrich
XtalFluor-M®
Pricing and availability is not currently available.
Sigma-Aldrich
XtalFluor-E®
Pricing and availability is not currently available.