- Synthesis and crystal structures of novel copper(II) complexes with glycine and substituted phenanthrolines: reactivity towards DNA/BSA and in vitro cytotoxic and antimicrobial evaluation.
Synthesis and crystal structures of novel copper(II) complexes with glycine and substituted phenanthrolines: reactivity towards DNA/BSA and in vitro cytotoxic and antimicrobial evaluation.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2016-11-11)
Duygu İnci, Rahmiye Aydın, Özgür Vatan, Tuba Sevgi, Dilek Yılmaz, Yunus Zorlu, Yusuf Yerli, Bünyemin Çoşut, Elif Demirkan, Nilüfer Çinkılıç
PMID27830402
ABSTRACT
New copper(II) complexes-dimeric-[Cu(nphen)(gly)(H