Merck
  • Synthesis, Spectral investigation (¹H, ¹³C) and Anti-microbial Screening of benzophenone imines.
Synthesis, Spectral investigation (¹H, ¹³C) and Anti-microbial Screening of benzophenone imines.
Pakistan journal of pharmaceutical sciences (2015-12-08)
Muhammad Kaleem Khosa, Muhammad Asghar Jamal, Muhammad Jawad Saif, Majid Muneer, Fazalur Rehman, Muhammad Farman, Hafiz Muhammad Shoaib, Muhammad Shahid, Shabnam Hameed
ABSTRACT
New series of benzophenone imines with general formula Ph2-C=NR; R = Benzyl, 4-Fluorobenzyl, Naphthyl, Phenyl, 4-Nitrophenyl were synthesized by condensation of dichlorodiphenylmethane and different aromatic primary amines (1:1) Those imines were characterized by different physiochemical and spectroscopic techniques like melting point, elemental analysis, FT-IR, multinuclear NMR (¹H, ¹³C). After characterization, imines were subjected to anti-microbial activities. All compounds showed promising activity against different bacterial strains like Escherichia coli, Bacillussubtilis, Pasturellam ultocida and Staphylococcus aureus as well as fungal strains like Alternata alternaria, Ganoderma lucidium, Penicillium notatum and Trichoderma harzianum using Amoxicillin and Flucanazole as a standard drugs respectively.
Sigma-Aldrich
4-Fluorobenzylamine, 97%