Merck
  • Carbazole hydroxylation by the filamentous fungi of the Cunninghamella species.
Carbazole hydroxylation by the filamentous fungi of the Cunninghamella species.
Environmental science and pollution research international (2015-08-16)
K Zawadzka, P Bernat, A Felczak, K Lisowska
ABSTRACT
Nitrogen heterocyclic compounds, especially carbazole, quinolone, and pyridine are common types of environmental pollutants. Carbazole has a toxic influence on living organisms, and the knowledge of its persistence and bioconversion in ecosystems is still not complete. There is an increasing interest in detoxification of hazardous xenobiotics by microorganisms. In this study, the ability of three filamentous fungi of the Cunninghamella species to eliminate carbazole was evaluated. The Cunninghamella elegans IM 1785/21Gp and Cunninghamella echinulata IM 2611 strains efficiently removed carbazole. The IM 1785/21Gp and IM 2611 strains converted 93 and 82 % of the initial concentration of the xenobiotic (200 mg L(-1)) after 120 h incubation. 2-Hydroxycarbazole was for the first time identified as a carbazole metabolite formed by the filamentous fungi of the Cunninghamella species. There was no increase in the toxicity of the postculture extracts toward Artemia franciscana. Moreover, we showed an influence of carbazole on the phospholipid composition of the cells of the tested filamentous fungi, which indicated its harmful effect on the fungal cell membrane. The most significant modification of phospholipid levels after the cultivation of filamentous fungi with the addition of carbazole was showed for IM 1785/21Gp strain.
Sigma-Aldrich
4-Hydroxycarbazole, 95%
Sigma-Aldrich
2-Hydroxycarbazole, 97%