Merck
RNAi-mediated gene function analysis in skin.
Methods in molecular biology (Clifton, N.J.) (2013-01-18)
Slobodan Beronja, Elaine Fuchs
ABSTRACT
We have recently developed a method for RNAi-mediated gene function analysis in skin (Beronja et al., Nat Med 16:821-827, 2010). It employs ultrasound-guided in utero microinjections of lentivirus into the amniotic cavity of embryonic day 9 mice, which result in rapid, efficient, and stable transduction into mouse skin. Our technique greatly extends the available molecular and genetic toolbox for comprehensive functional examination of outstanding problems in epidermal biology. In its simplest form, as a single-gene function analysis via shRNA-mediated gene knockdown, our technique requires no animal mating and may need as little as only a few days between manipulation and phenotypic analysis.
Sigma-Aldrich
Mineral oil, light oil, suitable for mouse embryo cell culture, BioXtra
Pricing and availability is not currently available.