Merck
Modified surface-active monomers for adhesive bonding to dentin.
Journal of dental research (1993-03-01)
S Venz, B Dickens
ABSTRACT
PMGDM, a PMDM-type adhesive monomer, was synthesized from pyromellitic dianhydride and glycerol dimethacrylate. Only the para isomer of PMDM, a solid, is used in dental adhesives. The adhesive monomer PMGDM is a liquid and consists of a mixture of para- and meta isomers. This study shows that PMGDM has several advantages over PMDM. For example, the adhesive can be applied at a greater thickness, and additives and/or additional initiators for improved curing can be incorporated. Use of the liquid monomer PMGDM also provided a convenient means of varying the thickness of the adhesive layer between dentin and composite. The adhesive bonds were tested in both shear and tensile modes. PMDM was used as the control. The reliability of the bonds, as judged from the Weibull modulus and Weibull characteristic strength, was improved by (1) use of more concentrated solutions of PMGDM adhesive, (2) use of an adhesive thickness of about 25 microns, and (3) modification of PMGDM with a diluent monomer which is expected to enhance the degree of cure and/or the dentin-wetting properties of the adhesive resin.
Sigma-Aldrich
mono-2-(Methacryloyloxy)ethyl maleate
Sigma-Aldrich
Pyromellitic dianhydride glycerol dimethacrylate, mixture of isomers, contains 1000-4000 ppm BHT as inhibitor
Sigma-Aldrich
N-Phenylglycine, 97%