Merck
  • [An arterial substance for subsequent injection during the preservation of the whole corpse].
[An arterial substance for subsequent injection during the preservation of the whole corpse].
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft (1992-06-01)
W Thiel
ABSTRACT
Masses for arterial injection were evaluated in 360 whole cadavers that had been injected with preservation fluid. A mass consisting of dextrin, latex, and lead tetroxide produced the best results. This mass provided high radiopacity at large distances from the injection site and ideal firmness for dissection. The easy application of the mass makes it suitable for research purposes as well as for teaching dissections.
Sigma-Aldrich
Lead(II,IV) oxide, ≥96%
Sigma-Aldrich
Lead oxide, powder (red), 1-2 μm, 99%