- Confirmation of leucogentian violet in chicken fat by gas chromatography/mass spectrometry.
Confirmation of leucogentian violet in chicken fat by gas chromatography/mass spectrometry.
A gas chromatographic/mass spectrometric (GC/MS) procedure for confirming the identity of leucogentian violet (LGV) in chicken fat was developed for regulatory application. The unused portion of the extract remaining from a determinative procedure was back-extracted into an organic phase, concentrated, and analyzed by GC/MS. Confirmation of the identity of LGV was based on matching the retention times and relative abundances of 6 ions in the extract to corresponding values obtained for the LGV standard. The procedure was validated by replicate analyses of negative control, fortified control, and residue-incurred chicken fat. The presence of LGV was confirmed by the GC/MS procedure in all samples found to contain LGV by prior liquid chromatographic analyses. There were no interferences in the control samples.