Merck
  • Preserving charge and oxidation state of Au(III) ions in an agent-functionalized nanocrystal model system.
Preserving charge and oxidation state of Au(III) ions in an agent-functionalized nanocrystal model system.
ACS nano (2011-07-09)
Stefan Müllegger, Wolfgang Schöfberger, Mohammad Rashidi, Thomas Lengauer, Florian Klappenberger, Katharina Diller, Kamuran Kara, Johannes V Barth, Eva Rauls, Wolf Gero Schmidt, Reinhold Koch
ABSTRACT
Supporting functional molecules on crystal facets is an established technique in nanotechnology. To preserve the original activity of ionic metallorganic agents on a supporting template, conservation of the charge and oxidation state of the active center is indispensable. We present a model system of a metallorganic agent that, indeed, fulfills this design criterion on a technologically relevant metal support with potential impact on Au(III)-porphyrin-functionalized nanoparticles for an improved anticancer-drug delivery. Employing scanning tunneling microscopy and -spectroscopy in combination with photoemission spectroscopy, we clarify at the single-molecule level the underlying mechanisms of this exceptional adsorption mode. It is based on the balance between a high-energy oxidation state and an electrostatic screening-response of the surface (image charge). Modeling with first principles methods reveals submolecular details of the metal-ligand bonding interaction and completes the study by providing an illustrative electrostatic model relevant for ionic metalorganic agent molecules, in general.
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, 97%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, ≥99%