Merck
  • Accurate transcription initiation by RNA polymerase II in a soluble extract from isolated mammalian nuclei.
Accurate transcription initiation by RNA polymerase II in a soluble extract from isolated mammalian nuclei.
Nucleic acids research (1983-03-11)
J D Dignam, R M Lebovitz, R G Roeder
ABSTRACT
We have developed a procedure for preparing extracts from nuclei of human tissue culture cells that directs accurate transcription initiation in vitro from class II promoters. Conditions of extraction and assay have been optimized for maximum activity using the major late promoter of adenovirus 2. The extract also directs accurate transcription initiation from other adenovirus promoters and cellular promoters. The extract also directs accurate transcription initiation from class III promoters (tRNA and Ad 2 VA).
Sigma-Aldrich
HeLa Nuclear Extract
Sigma-Aldrich
NuCLEAR Extraction Kit, For mammalian tissue or cultured cells