Merck
  • Differentiation of mouse embryonic stem cells into cells with spermatogonia-like morphology with chemical intervention-dependent increased gene expression of LIM homeobox 1 (Lhx1).
Differentiation of mouse embryonic stem cells into cells with spermatogonia-like morphology with chemical intervention-dependent increased gene expression of LIM homeobox 1 (Lhx1).
Stem cell research (2022-04-09)
Cameron Moshfegh, Sebastian G Rambow, Seraina A Domenig, Aldona Pieńkowska-Schelling, Ulrich Bleul, Viola Vogel
ABSTRACT
Spermatogonial stem cells (SSCs) originate from gonocytes that differentiate from primordial germ cells (PGCs). In the developing mouse testis, expression of the gene LIM homeobox 1 (Lhx1) marks the most undifferentiated SSCs, which has not yet been reported for spermatogonia-like cells generated in vitro. Previously, it was shown that a chemical intervention in male mouse embryonic stem (ES) cells in serum culture, including Sirtuin 1 (SIRT1) inhibitor Ex-527, DNA methyltransferase (DNMT) inhibitor RG-108 and electrophilic redox cycling compound tert-butylhydroquinone (tBHQ), was associated with molecular markers of PGC to gonocyte differentiation. Here, we report the in vitro differentiation of male mouse ES cells, cultured under dual chemical inhibition of GSK3β and MEK (2i) with leukemia inhibitory factor (LIF) (2iL) and serum, into cells with spermatogonia-like morphology (CSMs) and population-averaged expression of spermatogonia-specific genes by removal of 2iL and a specific schedule of twice daily partial medium replacement. Combination of this new protocol with the previously reported chemical intervention increased population-averaged gene expression of Lhx1 in the resulting CSMs. Furthermore, we detected single CSMs with strong nuclear LHX1/5 protein signal only in the chemical intervention group. We propose that further investigation of CSMs may provide new insights into male germline development.