Merck
  • Use of High-Performance Liquid Chromatography-Mass Spectrometry (HPLC-MS) to Quantify Modified Nucleosides.
Use of High-Performance Liquid Chromatography-Mass Spectrometry (HPLC-MS) to Quantify Modified Nucleosides.
Methods in molecular biology (Clifton, N.J.) (2022-03-16)
Rebecca Rodell, Ning Tsao, Adit Ganguly, Nima Mosammaparast
ABSTRACT
Physiological and chemically induced modifications to nucleosides are common in both DNA and RNA. Physiological forms of these modifications play critical roles in gene expression, yet aberrant marks, if left unrepaired, may be associated with increased genome instability. Due to the low prevalence of these marks in most samples of interest, a highly sensitive method is needed for their detection and quantitation. High-performance liquid chromatography, coupled to mass spectrometry (HPLC-MS), provides this high degree of sensitivity while also being adaptable to nearly any modified nucleoside of interest and still maintaining exquisite specificity. In this chapter, we demonstrate how to use HPLC-MS to analyze the catalytic activity of a nucleic acid demethylase, to quantify the prevalence of N6-methyladenosine from RNA, and to determine the kinetics of alkylation damage repair.
Sigma-Aldrich
Nuclease S1 from Aspergillus oryzae, for single-strand DNA/RNA digestion
Pricing and availability is not currently available.