Merck
Mammalian DNMTs in the male germ line DNA of Drosophila.
Biochemistry and cell biology = Biochimie et biologie cellulaire (2008-10-17)
Alexandra Weyrich, Xiaojing Tang, Guoliang Xu, André Schrattenholz, Christian Hunzinger, Wolfgang Hennig
ABSTRACT
It is controversial whether DNA methylation plays a functional role in Drosophila. We have studied testis DNA of Drosophila melanogaster Meigen, 1830 with antisera against 5-methylcytosine (5mC) and found no evidence for the presence of significant amounts of 5mC. Reactions occur only with 1 of 3 5mC antisera, but they are restricted to nuclear regions without detectable amounts of DNA. The antisera apparently cross-react with other nuclear components. If the murine de novo DNA methyltransferases, DNMT3A and DNMT3B, are expressed under the control of the spermatocyte-specific beta2-tubulin promoter in testes, DNA methylation is not increased and no effects on the fertility of the fly are seen. DNA methylation has, therefore, no functional relevance in the male germ line of Drosophila.
Sigma-Aldrich
Leupeptin trifluoroacetate salt, ≥90% (HPLC), microbial