Merck
  • Interaction of spirodiclofen with insecticides for the control of Brevipalpus yothersi in citrus.
Interaction of spirodiclofen with insecticides for the control of Brevipalpus yothersi in citrus.
Pest management science (2018-03-25)
Jaqueline F Della Vechia, Marcelo C Ferreira, Daniel J Andrade
ABSTRACT
The mite Brevipalpus yothersi (Acari: Tenuipalpidae) is of great importance in citriculture because it is a vector of the disease citrus leprosis. The frequency of application of a combination of pesticides has increased because of an increase in the occurrence of economically important pests of citrus. However, the combined effect of acaricide and other insecticides targeting B. yothersi is unknown. The objective of this study was to evaluate the effect of a combination of spirodiclofen and other insecticides on the control of B. yothersi. Analysis using spirodiclofen alone resulted in 97.22% efficiency in the control of B. yothersi on day 7 after application. A combination of spirodiclofen with either phosmet or imidacloprid resulted in 55% and 59% efficiency, respectively. A reduction in the efficiency of mite control by up to 42% was observed when the mites were exposed to a combination of acaricide with other insecticides. Application of a combination of spirodiclofen with phosmet or imidacloprid compromises the efficiency of acaricide in the control of B. yothersi. © 2018 Society of Chemical Industry.
Product Description
Supelco
Spirodiclofen, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland